Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIT, Tocris Bioscience™

Catalog No. 148010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-801-0 10 mg
14-805-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-801-0 Supplier R&D Systems Supplier No. 1480/10
This item is not returnable. View return policy

Irreversible δ agonist

Selective and irreversible δ-opioid agonist (EC50 = 8 nM); acts by alkylating the receptor.

Chemical Identifiers

CAS 85951-63-9
Molecular Formula C23H27N3OS
Molecular Weight (g/mol) 393.549
InChI Key VDKBIFPJULZUPU-UHFFFAOYSA-N
Synonym fentanyl isothiocyanate, fit, fentanyl ncs, n-1-2-4-isothiocyanatophenyl ethyl piperidin-4-yl-n-phenylpropanamide, fit alkylating agent, tocris-1480, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenylpropanamide, n-1-2-4-isothiocyanato-phenyl-ethyl-piperidin-4-yl-n-phenyl-propionamide, propanamide, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenyl
PubChem CID 84008
IUPAC Name N-[1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl]-N-phenylpropanamide
SMILES CCC(=O)N(C1CCN(CC1)CCC2=CC=C(C=C2)N=C=S)C3=CC=CC=C3

Specifications

Quantity 10 mg
Formula Weight 393.55
Percent Purity >99%
Chemical Name or Material FIT
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.