Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Antibody (2A3cc), Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $426.00
Specifications
Antigen | FITC |
---|---|
Clone | 2A3cc |
Dilution | Immunohistochemistry, Cell depletion |
Applications | Immunohistochemistry, Depletion Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1107898SS
![]() |
Novus Biologicals
NB1107898SS |
0.1 mg |
Each for $207.00
|
|
|||||
NB1107898
![]() |
Novus Biologicals
NB1107898 |
0.5 mg |
Each for $426.00
|
|
|||||
Description
FITC Monoclonal specifically detects FITC in Non-species specific samples. It is validated for Western Blot, ELISA, Sandwich ELISA, Cell depletion.Specifications
FITC | |
Immunohistochemistry, Cell depletion | |
Monoclonal | |
Purified | |
RUO | |
Fluorescein Isothiocyanate | |
IgG1 | |
Protein A purified | |
This product reacts with both free fluorescein (FITC) and conjugated FITC, so it recognizes FITC-labeled proteins. Kd = 1.5 x 10^-8 M |
2A3cc | |
Immunohistochemistry, Depletion Assay | |
Unconjugated | |
Mouse | |
Non-species specific | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with FITC-BSA conjugate. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title