Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Antibody - Azide and BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10064617
Description
FITC Polyclonal specifically detects FITC in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.Specifications
| FITC | |
| Polyclonal | |
| Unconjugated | |
| 0.1M BBS (pH 8.2) with No Preservative | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| Recognizes Fluorescein isothiocyanate. This preparation has been cross-absorbed against mouse, human and rat serum proteins. | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Fluorophore-linked immunosorbent assay | |
| Fluorescein Isothiocyanate | |
| Fluorescein isothiocyanate (isomer 1) | |
| 1 mg | |
| Primary | |
| Other | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction