Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Antibody, DyLight 594 , Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NB10064617DL594
Description
FITC Polyclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, ELISASpecifications
FITC | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Immunogen affinity purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
DyLight 594 | |
Fluorescein Isothiocyanate | |
Pooled antisera from sheep hyperimmunized with fluorescein isothiocyanate (Isomer I) | |
1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction