Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. G148C
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G148C 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. G148C Supplier R&D Systems Supplier No. G148C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FITC Polyclonal specifically detects FITC in Multi-Species samples. It is validated for .

Specifications

Antigen FITC
Classification Polyclonal
Conjugate Unconjugated
Dilution Hidden
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative
Gene Alias Fluorescein Isothiocyanate
Host Species Goat
Immunogen FITC-KLH (Fluorescein-isothiocyanate-coupled Keyhole Limpet Hemocyanin)
Purification Method Antigen Affinity-purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Fluorescein Isothiocyanate (FITC) in direct ELISAs. In direct ELISAs, nocross-reactivity with Alexa Fluor™ 488, Alexa Fluor™ 594, Alexa Fluor™ 647,Alexa Fluor™ 700, Alexa Fluor™ 750, NorthernLights™ NL493, NorthernLights™ 557,and NorthernLights™ 637 is observed
Reconstitution Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 0.2 mg/mL
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.