Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Antibody (SPM395), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308492MFV500
Description
FITC Monoclonal antibody specifically detects FITC in Non-species specific samples. It is validated for Flow CytometrySpecifications
FITC | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Non-species specific | |
Purified |
Flow Cytometry | |
SPM395 | |
Flow Cytometry | |
Fluorescein Isothiocyanate | |
Fluorescein isothiocyanate (FITC) conjugated to KLH | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction