Learn More
Invitrogen™ FITC Monoclonal Antibody (1F8-1E4), TRITC

Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Fluorophore: Tetramethyl rodamine 5 (and 6)-isothiocyanate (TRITC) Amax = 550 nm; Emax = 570 nm Fluorophore/Protein Absorbance Ratio: A550/A280 = 0.44 Product #31687 is recommended for use in ELISA, immunocytochemical, immunofluorescent, immunohistochemical, immunoprecipitation and Western blot applications. Restoration and Storage: Store product at 4°C until opened. Restore with 0.35 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for up to 6 weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of 50% glycerol and store at -20°C; or as an alternative, aliquot without glycerol and store at -80°or below, avoiding repeated freezing and thawing. Country of Origin: USA.
Specifications
Specifications
| Antigen | FITC |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1F8-1E4 |
| Concentration | 1.7 mg/mL |
| Conjugate | TRITC |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Gene Alias | Fluorescein |
| Host Species | Mouse |
| Purification Method | Purified |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.