Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Mouse anti-All, Clone: SPM395, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen FITC
Clone SPM395
Concentration 0.2 mg/ml
Dilution Western Blot, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, In-situ Hybridization
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123030
SDP
View Documents
Novus Biologicals
NBP30777820UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

FITC Monoclonal specifically detects FITC in All Species samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In-situ Hybridization.
Specifications

Specifications

FITC
0.2 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Unconjugated
Mouse
All species
Fluorescein Isothiocyanate
IgG1 κ
Protein A or G purified
SPM395
Western Blot, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, In-situ Hybridization
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Fluorescein isothiocyanate (FITC) conjugated to KLH
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.