Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FITC Mouse Anti-Human HLA-A3, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570463
Description
HLA-A3 Monoclonal antibody specifically detects HLA-A3 in Human samples. It is validated for Flow cytometry
Specifications
HLA-A3 | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
GAP.A3 (also known as GAP A3, GAP-A3) | |
FITC | |
HLA class I histocompatibility antigen, A-3 alpha chain; HLA Antigen A3 | |
Human Lymphocytes from Mixed Lymphocyte Reaction | |
25 μg | |
3105 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction