Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC Mouse, Clone: SPM395, Abnova™

Catalog No. 89035115 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-115 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-115 Supplier Abnova Corporation Supplier No. MAB15095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against FITC.

  • High specificity
  • High reactivity

Specifications

Antigen FITC
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone SPM395
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against FITC.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) In-Situ Hybridization Western Blotting The optimal working dilution should be
Formulation In 10 mM PBS.
Host Species Mouse
Immunogen FITC-KLH conjugated.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.