Learn More
Invitrogen™ FITC Polyclonal Antibody

Description
Rabbit anti FITC antibody recognizes fluorescein and does not react with immunoglobulins from any species.
Specifications
Specifications
| Antigen | FITC |
| Applications | ELISA, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene Alias | Fluorescein |
| Host Species | Rabbit |
| Immunogen | FITC Isomer 1. |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.