Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ FITC Protein Blocking Agent (PBA)

Catalog No. ta006pba
Click to view available options
Quantity:
6 mL

Block nonspecific antibody binding of primary reagent with Epredia™ FITC Protein Blocking Agent (PBA)

  • For In Vitro Diagnostic use
  • Blocks nonspecific antibody (protein) binding of primary reagent

Storage Conditions: Store at 2°-8°C.

Recommended for: Fluorescence Microscopy.

Specifications

Capacity 6 mL
Content And Storage 2° to 8°C.
Quantity 6 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.