Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FITM
GREENER_CHOICE

Catalog No. 685910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-591-0 10 mg
68-595-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-591-0 Supplier Tocris Bioscience™ Supplier No. 6859/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective negative allosteric modulator of mGlu1 receptors

FITM is a potent and selective negative allosteric modulator of mGlu1 receptors (IC50 = 5.1 nM). Displays >1300-fold selectivity for mGlu1 over other metabotropic glutamate receptors (IC50 values are 7 μM and >10 μM for mGlu5 and mGlu2 or mGlu8, respectively). Antagonizes methamphetamine-induced locomotion in mice.

Specifications

Quantity 10 mg
Molecular Formula C18H18FN5OS
Formula Weight 371.43
Product Type Negative Allosteric Modulator
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.