Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITM1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA554094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554094 Supplier Thermo Scientific Supplier No. PA554094

Rabbit Polyclonal Antibody

Immunogen sequence: RRVAVTARHL SRLVVGAAVW RGAGRAFLLI EDLTGSCFEP LPQGLLLHEL PDRRSCLAAG HQWRGYTVSS HTF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The specific function of this protein remains unknown.FIT1 belongs to an evolutionarily conserved family of proteins involved in fat storage (Kadereit et al., 2008 [PubMed 18160536]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FITM1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FITM1
Gene Accession No. A5D6W6
Gene Alias 1110028A07Rik; AV010547; Cg10671; fat storage inducing transmembrane protein 1; fat storage-inducing transmembrane protein 1; Fat-inducing protein 1; fat-inducing transcript 1; FIT1; FITM1; LOC161247; RGD1306911
Gene Symbols FITM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FITM1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.