Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FITM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544060 Supplier Invitrogen™ Supplier No. PA544060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MERGPVVGAG LGAGARIQAL LGCLLKVLLW VASALLYFGS EQAARLLGSP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 92%; Rat: 86%.

The specific function of this protein remains unknown.FIT1 belongs to an evolutionarily conserved family of proteins involved in fat storage (Kadereit et al., 2008 [PubMed 18160536]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FITM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FITM1
Gene Accession No. A5D6W6
Gene Alias 1110028A07Rik; AV010547; Cg10671; fat storage inducing transmembrane protein 1; fat storage-inducing transmembrane protein 1; Fat-inducing protein 1; fat-inducing transcript 1; FIT1; FITM1; LOC161247; RGD1306911
Gene Symbols FITM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FITM1 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161247
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.