Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIV Envelope Antibody (CE4-13B1 (13B1)) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | FIV Envelope |
---|---|
Clone | CE4-13B1 (13B1) |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunocytochemistry, Western Blot, ELISA, Immunofluorescence, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017784
![]() |
Novus Biologicals
NB100665530.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB100665531
![]() |
Novus Biologicals
NB100665530.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
FIV Envelope Monoclonal specifically detects FIV Envelope in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FIV Envelope | |
1.0 mg/mL | |
Immunocytochemistry, Western Blot, ELISA, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Mouse | |
Virus | |
Purified gp95 envelope antigen. | |
Primary | |
Protein A purified | |
NB100-66553 is specific for FIV Envelope gp95 protein. This antibody is directed against an epitope within the variable V2 region and cross-reacts with Petaluma, 34TF10, Ppr, MCH5-5 and Glasgow 8 isolates. NB100-66553 does not neutralize FIV and does not recognize Feline Leukaemia Virus, Feline Herpes Virus Type 1, Feline Coronavirus and Feline Calicivirus. |
CE4-13B1 (13B1) | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) with 0.05% Sodium Azide | |
IgG2a | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title