Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIV Envelope Antibody (CE4-13B1 (13B1)), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066553V0.1ML
Description
FIV Envelope Monoclonal specifically detects FIV Envelope in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FIV Envelope | |
Monoclonal | |
DyLight 405 | |
50 mM sodium borate with 0.05% sodium azide | |
Purified gp95 envelope antigen. | |
Primary | |
Virus | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
CE4-13B1 (13B1) | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
0.1 mL | |
NB100-66553 is specific for FIV Envelope gp95 protein. This antibody is directed against an epitope within the variable V2 region and cross-reacts with Petaluma, 34TF10, Ppr, MCH5-5 and Glasgow 8 isolates. NB100-66553 does not neutralize FIV and does not recognize Feline Leukaemia Virus, Feline Herpes Virus Type 1, Feline Coronavirus and Feline Calicivirus. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction