Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIV Envelope Antibody (CE4-13B1 (13B1)), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066553MFV610
Description
FIV Envelope Monoclonal antibody specifically detects FIV Envelope in Virus samples. It is validated for Flow CytometrySpecifications
FIV Envelope | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Purified gp95 envelope antigen. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
CE4-13B1 (13B1) | |
Flow Cytometry | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction