Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FIV gp41 Monoclonal Antibody (5A1)
GREENER_CHOICE

Catalog No. PIMA183167
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA183167 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA183167 Supplier Invitrogen™ Supplier No. MA183167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot, this antibody detects a band at ~140 and 41 kDa under reducing conditions. Mouse anti Feline immunodeficiency virus gp41 transmembrane protein antibody, clone 5A1 recognizes feline immunodeficiency virus transmembrane protein gp41.

The lentivirus FIV is responsible for a progressive and debilitating immunodeficiency syndrome in domestic cats, similar to that caused by HIV. It is a complex retrovirus with a tightly-packed genome containing the structrual genes gag, env and pro-pol, as well as accessory genes vif, rev and ORF-A/2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FIV gp41
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene Alias Feline Immunodeficiency Virus
Host Species Mouse
Immunogen Native, FIV extracts.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.