Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIV matrix protein p15Rabbit anti-Virus, Polyclonal, Rockland™
SDP

Catalog No. 501991906
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1906 Supplier Rockland Immunochemicals Supplier No. 600401992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against FIV Matrix Protein p15.

The Gag family is characterized by assembly of the viral capsid at either the plasma membrane or at the limiting membrane of late endosomes. The capsid assembles from the viral Gag polyprotein. Upon release of a budding virion, Gag precursor protein is cleaved by the viral protease into its mature products, namely Matrix Protein, Capsid and Nucleocapsid. Matrix Protein, located at the N-terminus of the Gag polyprotein, is usually myristylated during protein translation, prior to the later events of virus assembly. The myristate moiety is believed to be sequestered within the Matrix Protein during protein translation and later facilitates membrane binding upon exposure resulting from conformational changes. Essential functions attributed to the Matrix Protein of lentiviruses include targeting newly synthesized Gag precursor proteins to the site of virus assembly by binding with cellular components such as phosphatidylinositides. In the mature virus particle, the Matrix Protein provides internal structure to the virion within the capsid, but is not exposed at the surface of the particle. Based on studies with HIV, it is postulated that FIV Matrix Protein may also serve additional functions, including nuclear localization of the viral core upon entry of the virus into a new host cell.

Specifications

Antigen FIV matrix protein p15
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.