Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 455UV

Catalog No. 501128846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 tests
500 tests
Catalog No. Quantity
50-112-8846 100 tests
50-112-8879 500 tests
2 options

Catalog No. 50-112-8846

Supplier: Invitrogen™ 65086814

Only null left
Add to Cart
Edge
Add to Cart

Can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and PI, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized and stained for intracellular antigens without loss of staining intensity.

  • Fixable Viability Dyes may be used to label cells from all species
  • Fixable Viability Dye eFluor™ 455UV can be excited by a UV laser line (≈350nm) and has a peak emission of 455nm
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • Fixable Viability Dye eFluor™ 455UV is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture
  • Store at −70°C with dessicant
  • It may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
  • Staining may be done before or after surface staining
  • Cells may be cryopreserved after staining with no adverse effect on staining intensity of dead cells after thawing
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
Emission 350/455
Product Line eBioscience
Quantity 100 tests
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.