Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIZZ1/RELM alpha Antibody (RM0313-9A39) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | FIZZ1/RELM alpha |
---|---|
Clone | RM0313-9A39 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Monoclonal |
Description
FIZZ1/RELM alpha Monoclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FIZZ1/RELM alpha | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q9EP95 | |
57262 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0313-9A39 | |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
1810019L16Rik, Fizz1, Fizz-1, HIMF, RELMa, RELMalpha, RELM-alpha, Retnla resistin like alpha, Xcp2 | |
RETNLA | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title