Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse FIZZ1/RELM alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$214.00 - $2020.00
Specifications
Antigen | FIZZ1/RELM alpha |
---|---|
Clone | 226033 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1523SP
![]() |
R&D Systems
MAB1523SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1523100
![]() |
R&D Systems
MAB1523100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1523500
![]() |
R&D Systems
MAB1523500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
FIZZ1/RELM alpha Monoclonal specifically detects FIZZ1/RELM alpha in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FIZZ1/RELM alpha | |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q9EP95 | |
57262 | |
E. coli-derived recombinant mouse FIZZ1/RELM alpha Asp24-Ser111 Accession # Q9EP95 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse FIZZ1/RELM alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) RELM beta or rmResistin is observed. |
226033 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
1810019L16Rik, Fizz1, Fizz-1, HIMF, RELM alpha, RELMa, RELMalpha, RELM-alpha, RETNLA, Retnla resistin like alpha, RETNLB, Xcp2 | |
RETNLA | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title