Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FJX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597198 Supplier Invitrogen™ Supplier No. PA597198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAQAAAWLEA ARGARMVALE RGGCGRSSNR LARFADGTRA CVRYGINPEQ IQGEALSYYL ARLLGLQRHV PPLALARVEA RGAQWAQVQE ELRAAHWTEG SVVSLTRWLP NLTDVVVPAP WRSEDGRLRP LRDAGGELAN LSQAELVDLV QWTDLILFDY LTANFDRLVS NLFSLQWDPR VMQRATSNLH RGPGGALVFL DNEAGLVHGY RVAGMWDKYN EPLLQSVCVF RERTARRVLE LHRGQDAAAR LLRLYRRHEP RFPELAALAD PHAQLLQRRL DFLAKHILHC KAKYGRRSGT; Positive Samples: 293T, Rat kidney.

FJX1 is the human ortholog of mouse and Drosophila four-jointed gene product. The Drosophila protein is important for growth and differentiation of legs and wings, and for proper development of the eyes. The exact function of FJX1 gene in humans is not known..The protein encoded by this gene is the human ortholog of mouse and Drosophila four-jointed gene product. The Drosophila protein is important for growth and differentiation of legs and wings, and for proper development of the eyes. The exact function of this gene in humans is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FJX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FJX1
Gene Accession No. Q86VR8
Gene Alias FJX1; FLJ22416; FLJ25593; four jointed box 1; four jointed box 1 (Drosophila); four-jointed box kinase 1; four-jointed box protein 1; Four-jointed protein homolog; four-jointed protein x1; LOW QUALITY PROTEIN: four-jointed box protein 1; putative secreted ligand homologous to fjx1
Gene Symbols FJX1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 138-437 of human FJX1 (NP_0551592).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24147, 366140
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.