Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FK 506
GREENER_CHOICESDP

Catalog No. 363110R Shop All R&D Systems Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
36-311-0R 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 36-311-0R Supplier Tocris Bioscience™ Supplier No. 3631/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent calcineurin inhibitor; immunosuppressant

FK 506 is a potent calcineurin (protein phosphatase 2B) inhibitor that requires FK 506-binding protein 12 (FKBP12) for activity (IC50 = 3 nM). Inhibits secretion of IL-1, IL-2 (IC50 = 1 nM), IL-3, IL-4, IL-6 (IC50 = 35 nM), GM-CSF, TNFα (IC50 = 10 nM), IFNγ and Myc from activated T cells in vitro. Exhibits potent immunosuppressive, neuroprotective and anticonvulsant activity in vivo. Identified as targeting human host cell proteins that interact with SARS-CoV-2.

Analog also available.

Specifications

Quantity 10 mg
Product Type FK 506
Purity >99%
Molecular Weight (g/mol) 804.02
Inhibitors FK 506

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.