Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FK506 Antibody (FK1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100625710.025MG
Description
FK506 Monoclonal specifically detects FK506 in Non-species specific samples. It is validated for ELISA.Specifications
FK506 | |
Monoclonal | |
1 mg/mL | |
ELISA 1:100-1:2000 | |
Mouse | |
IgM purified | |
RUO | |
NB100-62571 detects FK-506, a macrolide with similar but more potent immunosuppressant activity than cyclosporine. It is a 23-membered lactone produced by the bacteria Streptomyces tsukubaensis. FK-506 inhibits signal transduction and interleukin-2 transcription in CD4+ T helper lymphocytes, and is mainly used after allogenic organ transplantation to reduce organ rejection. Side effects of FK-506 treatment include nephrotoxicity, neurotoxicity, diabetes mellitus and hypertension. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM |
ELISA | |
FK1 | |
Unconjugated | |
TBS with 0.09% Sodium Azide | |
FK-506-BSA conjugate | |
0.025 mg | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction