Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563387 Supplier Invitrogen™ Supplier No. PA563387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLPTGYLFVW HKDPPANLFE DMDLNKDGEV PPEEFSTFIK AQVSEGKGRL MPGQDPEKTI GDMFQNQDRN QDGKITV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

MAF acts as a transcriptional activator or repressor. The protein is involved in embryonic lens fiber cell development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FKBP10
Gene Accession No. Q96AY3
Gene Alias 65 kDa FK506-binding protein; 65 kDa FKBP; AI325255; BRKS1; FK506 binding protein 10; FK506 binding protein 10, 65 kDa; FK506 binding protein 6 (65 kDa); FK506-binding protein 10; FKBP10; FKBP-10; Fkbp1-rs; Fkbp6; FKBP65; FKBP-65; Fkbprp; Fkbp-rs; Fkbp-rs1; hFKBP65; Immunophilin FKBP65; OI11; OI6; Peptidyl-prolyl cis-trans isomerase FKBP10; PPIASE; PPIase FKBP10; PSEC0056; Rotamase
Gene Symbols FKBP10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FKBP10. Recombinant protein control fragment (Product #RP-103859).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.