Learn More
FKBP10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305516AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.Specifications
FKBP10 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q61576, Q96AY3 | |
FKBP10 | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FKBP10 | |
FK506-binding protein 10; Peptidyl-prolyl cis-trans isomerase; PPIase; Rotamase; Immunophilin FKBP65; 65 kDa FK506-binding protein; FKBP65; PSEC0056; FKBP65; FKBP10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14230, 60681 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.