Learn More
Invitrogen™ FKBP11 Polyclonal Antibody

Description
Immunogen sequence: EAGLETESPV RTLQVETLVE PPEPCAEPAA FGDTLHIHYT GSLVDGRIID TSLTRDPLVI ELGHKQVIPG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Specifications
Specifications
| Antigen | FKBP11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FKBP11 |
| Gene Accession No. | Q9D1M7, Q9NYL4 |
| Gene Alias | 1110002O23Rik; 19 kDa FK506-binding protein; 19 kDa FKBP; FK506 binding protein 11; FK506 binding protein 11, 19 kDa; FK506-binding protein 11; FKBP11; FKBP-11; FKBP19; FKBP-19; peptidyl-prolyl cis-trans isomerase FKBP11; PPIase FKBP11; rotamase; UNQ336/PRO535 |
| Gene Symbols | FKBP11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.