Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113417
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113417 Supplier Invitrogen™ Supplier No. PA5113417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP11 Polyclonal Antibody for ICC/IF, ELISA

PPIases accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP11
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FKBP11
Gene Accession No. Q9NYL4
Gene Alias 1110002O23Rik; 19 kDa FK506-binding protein; 19 kDa FKBP; FK506 binding protein 11; FK506 binding protein 11, 19 kDa; FK506-binding protein 11; FKBP11; FKBP-11; FKBP19; FKBP-19; peptidyl-prolyl cis-trans isomerase FKBP11; PPIase FKBP11; rotamase; UNQ336/PRO535
Gene Symbols FKBP11
Host Species Rabbit
Immunogen Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP11 protein (28-154AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51303
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.