Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP11 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13176R

Description
PPIases accelerate the folding of proteins during protein synthesis.Specifications
FKBP11 | |
Polyclonal | |
Unconjugated | |
FKBP11 | |
1110002O23Rik; 19 kDa FK506-binding protein; 19 kDa FKBP; FK506 binding protein 11; FK506 binding protein 11, 19 kDa; FK506-binding protein 11; FKBP11; FKBP-11; FKBP19; FKBP-19; Peptidyl-prolyl cis-trans isomerase FKBP11; PPIase FKBP11; Rotamase; UNQ336/PRO535 | |
Rabbit | |
Protein A | |
RUO | |
51303 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NYL4 | |
FKBP11 | |
KLH conjugated synthetic peptide derived from human FKBP11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction