Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP12.6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP302968100UL
Description
FKBP12.6 Polyclonal specifically detects FKBP12.6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FKBP12.6 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
12.6 kDa FK506-binding protein, calstabin 2,12.6 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1B, 12.6 kDa, FK506-binding protein 12.6, FK506-binding protein 1B, FK506-binding protein 1B (12.6 kD), FKBP-12.6, FKBP12.6h-FKBP-12, FKBP-1B, FKBP1L, FKBP9, Immunophilin FKBP12.6, OTK4PPIase FKBP1B, peptidyl-prolyl cis-trans isomerase FKBP1B, PKBP1L, PPIase, Rotamase | |
A synthetic peptide corresponding to a sequence within amino acids 1-80 of human FKBP12.6 (NP_473374.1). | |
100 μL | |
mTOR Pathway, Stem Cell Markers | |
2281 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction