Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP12 Antibody (OTI3B3), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270741MFV610
Description
FKBP12 Monoclonal antibody specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
FKBP12 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI3B3 | |
Flow Cytometry | |
12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
Full length human recombinant protein of human FKBP1A (NP_000792) produced in HEK293T cell. | |
0.1 mL | |
Autophagy, Cancer, mTOR Pathway | |
2280 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction