Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP12 Monoclonal Antibody (2137CT1015.65.47)
GREENER_CHOICE

Catalog No. PIMA537794
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537794 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537794 Supplier Thermo Scientific Supplier No. MA537794
Add to Cart
Add to Cart

FKBP12 Monoclonal Antibody (2137CT1015.65.47)

FKBP12 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Western Blot
Classification Monoclonal
Clone 2137CT1015.65.47
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FKBP1A
Gene Accession No. P26883, P62942, Q62658
Gene Alias 12 kDa FK506-binding protein; 12 kDa FKBP; calstabin 1; calstabin-1; EC 5.2.1.8; FK506 binding protein 1a; FK506 binding protein 1A, 12kDa; FK506 binding protein 2 (13 kDa); FK506 binding protein12; FK506-binding protein 1; FK506-binding protein 1 (12kD); FK506-binding protein 12; FK506-binding protein 1a; FK506-binding protein 1A (12kD); FK506-binding protein, T-cell, 12-kD; Fkbp; FKBP1; FKBP12; FKBP-12; FKBP12-Exip3; Fkbp1a; FKBP-1A; Fkbp2; Immunophilin FKBP12; peptidyl-prolyl cis-trans isomerase FKBP1A; PKC12; PKCI2; PPIASE; PPIase FKBP1A; protein kinase C inhibitor 2; rotamase
Gene Symbols FKBP1A
Host Species Mouse
Immunogen KLH conjugated synthetic peptide.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14225, 2280, 25639
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.