Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP12 Rabbit anti-Human, Mouse, Rat, Clone: 9A2J5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | FKBP12 |
---|---|
Clone | 9A2J5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
FKBP12 Monoclonal antibody specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
FKBP12 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, mTOR Pathway | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2280 | |
IgG | |
Affinity purified |
9A2J5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
A synthetic peptide corresponding to a sequence within amino acids 29-108 of human FKBP12 (P62942). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title