Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP13/FKBP2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297373MFV610
Description
FKBP13/FKBP2 Polyclonal antibody specifically detects FKBP13/FKBP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FKBP13/FKBP2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 5.2.1.8,13 kDa FK506-binding protein, FK506 binding protein 2, 13kDa, FK506-binding protein 2, FK506-binding protein 2 (13kD), FKBP13,13 kDa FKBP, FKBP-13proline isomerase, FKBP-2, Immunophilin FKBP13, peptidyl-prolyl cis-trans isomerase FKBP2, PPIase, PPIase FKBP2, rapamycin-binding protein, Rotamase | |
Produced in rabbits immunized with E. coli-derived Human FKBP13/FKBP2 fragment. | |
0.1 mL | |
Signal Transduction | |
2286 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction