Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP13/FKBP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23860425UL
Description
FKBP13/FKBP2 Polyclonal specifically detects FKBP13/FKBP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FKBP13/FKBP2 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
P26885 | |
FKBP2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RKLQIGVKKRVDHCPIKSRKGDVLHMHYT | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 5.2.1.8,13 kDa FK506-binding protein, FK506 binding protein 2, 13kDa, FK506-binding protein 2, FK506-binding protein 2 (13kD), FKBP13,13 kDa FKBP, FKBP-13proline isomerase, FKBP-2, Immunophilin FKBP13, peptidyl-prolyl cis-trans isomerase FKBP2, PPIase, PPIase FKBP2, rapamycin-binding protein, Rotamase | |
Rabbit | |
Affinity Purified | |
RUO | |
2286 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction