Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP14 Antibody (18E2), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259474APC
Description
FKBP14 Monoclonal antibody specifically detects FKBP14 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
FKBP14 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry | |
18E2 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
22 kDa FK506-binding protein, EC 5.2.1.8, FK506 binding protein 14 (22 kDa), FK506 binding protein 14, 22 kDa, FK506-binding protein 14, FKBP-14, FKBP-22, FKBP22PPIase FKBP14, FLJ20731, peptidyl-prolyl cis-trans isomerase FKBP14,22 kDa FKBP, rotamase | |
Recombinant human FKBP14 (20-211aa) purified from E. coli | |
0.1 mL | |
Developmental Biology | |
55033 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction