Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP14 Antibody (18E2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259474MFV500
Description
FKBP14 Monoclonal antibody specifically detects FKBP14 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FKBP14 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 18E2 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| 22 kDa FK506-binding protein, EC 5.2.1.8, FK506 binding protein 14 (22 kDa), FK506 binding protein 14, 22 kDa, FK506-binding protein 14, FKBP-14, FKBP-22, FKBP22PPIase FKBP14, FLJ20731, peptidyl-prolyl cis-trans isomerase FKBP14,22 kDa FKBP, rotamase | |
| Recombinant human FKBP14 (20-211aa) purified from E. coli | |
| 0.1 mL | |
| Developmental Biology | |
| 55033 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction