Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589090 Supplier Invitrogen™ Supplier No. PA589090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALIPEPEVKI EVLQKPFICH RKTKGGDLML VHYEGYLEKD GSLFHSTHKH NNGQPIWFTL GILEALKGWD QGLKGMCVGE KRKLIIPPAL GYGKEGKGKI PPESTLIFNI DLLEIRNGPR SHESFQEMDL NDDWKLSKDE VKAYLKKEFE KHGAVVNESH HDALVEDIFD KEDEDKDGFI SAREFTYKHD EL; Positive Samples: H460, HeLa, U-87MG, MCF-7, Mouse testis, Rat testis; Cellular Location: Endoplasmic reticulum lumen.

PPIases accelerate the folding of proteins during protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fkbp14
Gene Accession No. P59024, Q9NWM8
Gene Alias 22 kDa FK506-binding protein; 22 kDa FKBP; BC029109; EDSKMH; FK506 binding protein 14; FK506 binding protein 14, 22 kDa; FK506-binding protein 14; Fkbp14; FKBP-14; FKBP-2; FKBP22; FKBP-22; IPBP12; peptidyl-prolyl cis-trans isomerase FKBP14; PPIase; PPIase FKBP14; rotamase; UNQ322/PRO381
Gene Symbols Fkbp14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-211 of human FKBP14 (NP_0604161).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231997, 362366, 55033
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.