Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730552 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0552 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0552 Supplier Proteintech Group Inc Supplier No. 158841AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPIases accelerate the folding of proteins during protein synthesis.

Specifications

Antigen FKBP14
Applications Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FKBP14
Gene Accession No. P59024, Q9NWM8
Gene Alias 22 kDa FK506 binding protein, 22 kDa FKBP, FK506 binding protein 14, FKBP 14, FKBP 22, FKBP14, FKBP22, PPIase FKBP14, Rotamase
Gene Symbols Fkbp14
Host Species Rabbit
Immunogen FKBP14 Fusion Protein Ag8668
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231997, 362366, 55033
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.