Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552495
Description
Immunogen sequence: KHSAGNSMLI PSMSVTMETS MIMSNIQRII QENERLKQEI LEKSNRIEEQ NDKISELIER NQRYVEQSNL MMEKRNNSLQ TATENTQARV LHAEQEKAKV TEELAAATAQ VSHLQLKMTA HQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
FKBP15, also known as FKBP133, is a member of the FK506-binding protein family, a group of proteins initially identified as immunophilins, targets for the immunosupressant drugs FK506 and Rapamycin. FKBP15 is expressed in the developing nervous system and contains a domain similar to Wiskott-Aldrich syndrome protein homology region 1 (WH1) in addition to the FK506-binding protein motif. FKBP15 is distributed along the axonal shafts and partially co-localizes with F-actin in the growth cones of dorsal root ganglion neurons; overexpression of FKBP15 resulted in the number of filopodia in transfected neurons, suggesting that FKBP15 modulates growth cone behavior. FKBP15 has also been shown to associate with both microtubules and the actin filament systems and disruption of its expression by RNAi resulted in delayed transport of early endosomes in HeLa cells indicating that FKBP15 is also involved in the transport of early endosomes. At least three isoforms of FKBP15 are known to exist.
Specifications
| FKBP15 | |
| Polyclonal | |
| Unconjugated | |
| FKBP15 | |
| 133 kDa FK506-binding protein; 133 kDa FKBP; BB131447; C430014M02Rik; FK506 binding protein 15; FK506 binding protein 15, 133kDa; FK506-binding protein 133; FK506-binding protein 133kDa; FK506-binding protein 15; FKBO133; Fkbp133; FKBP-133; FKBP15; FKBP-15; Kiaa0674; mKIAA0674; PPP1R76; protein phosphatase 1, regulatory subunit 76; RGD1304816; WAFL; WASP and FKBP-like; WASP- and FKBP-like protein; WASP and FKBP-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23307 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T1M5 | |
| FKBP15 | |
| Recombinant protein corresponding to Human FKBP15. Recombinant protein control fragment (Product #RP-89625). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction