Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144010
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144010 Supplier Invitrogen™ Supplier No. PA5144010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hacat whole cell, human Raji whole cell, mouse lung tissue. IHC: human spleen tissue, human tonsil tissue, mouse brain tissue, rat brain tissue, human breast tissue, human lymphoma tissue. ICC/IF: MCF-7 cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

FKBP15, also known as FKBP133, is a member of the FK506-binding protein family, a group of proteins initially identified as immunophilins, targets for the immunosupressant drugs FK506 and Rapamycin. FKBP15 is expressed in the developing nervous system and contains a domain similar to Wiskott-Aldrich syndrome protein homology region 1 (WH1) in addition to the FK506-binding protein motif. FKBP15 is distributed along the axonal shafts and partially co-localizes with F-actin in the growth cones of dorsal root ganglion neurons; overexpression of FKBP15 resulted in the number of filopodia in transfected neurons, suggesting that FKBP15 modulates growth cone behavior. FKBP15 has also been shown to associate with both microtubules and the actin filament systems and disruption of its expression by RNAi resulted in delayed transport of early endosomes in HeLa cells indicating that FKBP15 is also involved in the transport of early endosomes. At least three isoforms of FKBP15 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP15
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene FKBP15
Gene Accession No. Q5T1M5, Q6P9Q6
Gene Alias 133 kDa FK506-binding protein; 133 kDa FKBP; BB131447; C430014M02Rik; FK506 binding protein 15; FK506 binding protein 15, 133kDa; FK506-binding protein 133; FK506-binding protein 133kDa; FK506-binding protein 15; FKBO133; Fkbp133; FKBP-133; FKBP15; FKBP-15; Kiaa0674; mKIAA0674; PPP1R76; protein phosphatase 1, regulatory subunit 76; RGD1304816; WAFL; WASP and FKBP-like; WASP- and FKBP-like protein; WASP and FKBP-like protein
Gene Symbols FKBP15
Host Species Rabbit
Immunogen E.coli-derived human FKBP135/FKBP15 recombinant protein (Position: A19-D1210).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23307, 338355, 362528
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.