Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583780 Supplier Invitrogen™ Supplier No. PA583780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RIGKQEVIKG FEEGAAQLGP LSPLPI Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 65%.

Has the potential to contribute to the immunosuppressive and toxic effects of FK506 and rapamycin. PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FKBP1B
Gene Accession No. P68106
Gene Alias 12.6 kDa FK506-binding protein; 12.6 kDa FKBP; 12.6kDa; 126 kDa FKBP; AW494148; calstabin 2; calstabin2; calstabin-2; FK506 binding protein 1B; FK506 binding protein 1B, 12.6 kDa; FK506-binding protein 12.6; FK506-binding protein 1B; FKBP12.6; FKBP-12.6; FKBP126; FKBP-126; FKBP1B; FKBP-1B; FKBP1L; FKBP9; h-FKBP-12; immunophilin FKBP12.6; OTK4; Peptidyl-prolyl cis-trans isomerase FKBP1B; PKBP1L; PPIase; PPIase FKBP1B; Rotamase
Gene Symbols FKBP1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FKBP1B. Recombinant protein control fragment (Product #RP-108367).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2281
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.