Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729991 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9991 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9991 Supplier Proteintech Group Inc Supplier No. 151141AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Has the potential to contribute to the immunosuppressive and toxic effects of FK506 and rapamycin. PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Specifications

Antigen FKBP1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FKBP1B
Gene Accession No. P68106, P97534, Q9Z2I2
Gene Alias 12.6 kDa FK506 binding protein, 12.6 kDa FKBP, FK506 binding protein 1B, FKBP 12.6, FKBP 1B, FKBP12.6, FKBP1B, FKBP1L, FKBP9, h FKBP 12, Immunophilin FKBP12.6, OTK4, PKBP1L, PPIase, PPIase FKBP1B, Rotamase
Gene Symbols Fkbp1b
Host Species Rabbit
Immunogen FKBP1B Fusion Protein Ag7153
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14226, 2281, 58950
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.