Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576346 Supplier Thermo Scientific Supplier No. PA576346
Add to Cart
Add to Cart

FKBP2 Polyclonal Antibody

FKBP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Fkbp2
Gene Accession No. P26885, P45878
Gene Alias 13 kDa FK506-binding protein; 13 kDa FKBP; 13kDa; FK506 binding protein 2; FK506 binding protein 2, 13kDa; FK506-binding protein 2; FK506-binding protein 2 (13kD); FKBP prolyl isomerase 2; FKBP13; FKBP-13; FKBP2; FKBP-2; Immunophilin FKBP13; mFKBP13; mFKBP2; Peptidyl-prolyl cis-trans isomerase FKBP2; PPIase; PPIase FKBP2; proline isomerase; rapamycin-binding protein; RGD1560660; Rotamase
Gene Symbols Fkbp2
Host Species Rabbit
Immunogen Recombinant protein of human FKBP2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14227, 2286, 293702
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.