Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FKBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117119 Supplier Invitrogen™ Supplier No. PA5117119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, SKOV3, Jurkat, HeLa, Mouse thymus, Mouse liver, Mouse brain Immunogen sequence: ATGAEGKRKL QIGVKKRVDH CPIKSRKGDV LHMHYTGKLE DGTEFDSSLP QNQPFVFSLG TGQVIKGWDQ GLLGMCEGEK RKLVIPSELG YGERGAPPKI PGGATLVFEV ELLKIERRTE L.

FKBP2 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin. It is thought to function as an ER chaperone and may also act as a component of membrane cytoskeletal scaffolds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fkbp2
Gene Accession No. P26885, P45878
Gene Alias 13 kDa FK506-binding protein; 13 kDa FKBP; 13kDa; FK506 binding protein 2; FK506 binding protein 2, 13kDa; FK506-binding protein 2; FK506-binding protein 2 (13kD); FKBP prolyl isomerase 2; FKBP13; FKBP-13; FKBP2; FKBP-2; Immunophilin FKBP13; mFKBP13; mFKBP2; Peptidyl-prolyl cis-trans isomerase FKBP2; PPIase; PPIase FKBP2; proline isomerase; rapamycin-binding protein; RGD1560660; Rotamase
Gene Symbols Fkbp2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-142 of human FKBP2 (NP_0044612).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14227, 2286, 293702
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.