Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP25 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP141071FR
Description
FKBP25 Polyclonal antibody specifically detects FKBP25 in Human samples. It is validated for Western BlotSpecifications
FKBP25 | |
Polyclonal | |
Western Blot | |
EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
The immunogen for this product maps to a region between residue 50 and 100 of human FK506 binding protein 3 using the numbering given in entry NP_002004.1 (GeneID 2287). | |
0.1 mL | |
Stem Cell Markers | |
2287 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction