Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FKBP25 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300022MFV450
Description
FKBP25 Polyclonal antibody specifically detects FKBP25 in Human samples. It is validated for ELISASpecifications
FKBP25 | |
Polyclonal | |
ELISA | |
EC 5.2.1.8,25 kDa FK506-binding protein, FK506 binding protein 3, 25kDa, FK506-binding protein 3, FK506-binding protein 3 (25kD), FKBP25,25 kDa FKBP, FKBP-3, Immunophilin FKBP25, peptidyl-prolyl cis-trans isomerase FKBP3, PPIase, PPIase FKBP3, rapamycin binding protein, Rapamycin-selective 25 kDa immunophilin, Rotamase, T-cell | |
Produced in rabbits immunized with purified, recombinant Human FKBP25 (Uniprot#: Q00688; Ala2-Asp224) | |
0.1 mL | |
Stem Cell Markers | |
2287 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction