Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551470
Description
Immunogen sequence: LPKKDIIKFL QEHGSDSFLA EHKLLGNIKN VAKTANKDHL VTAYNHLFET KRFKGTESIS KVSEQVKNVK LNEDKPKETK SEETLDEGPP KYTKSVLKKG DKTNFPKKGD VVHCWYTGTL QDGT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.
Specifications
| FKBP25 | |
| Polyclonal | |
| Unconjugated | |
| FKBP3 | |
| 25 kDa FK506-binding protein; 25 kDa FKBP; 25kDa; FK506 binding protein 3; FK506 binding protein 3, 25kDa; FK506-binding protein 25, T-cell; FK506-binding protein 3; FK506-binding protein 3 (25kD); FKBP25; FKBP-25; Fkbp3; FKBP-3; Immunophilin FKBP25; peptidyl-prolyl cis-trans isomerase FKBP3; PPIase; PPIase FKBP3; rapamycin binding protein; rapamycin-selective 25 kDa immunophilin; Rotamase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2287, 299104, 30795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q00688, Q62446 | |
| FKBP3 | |
| Recombinant protein corresponding to Human FKBP25. Recombinant protein control fragment (Product #RP-88925). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction